Product Details
- SNP ID
-
rs200591173
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:17816760 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGCCAGCAGTGGGGGCAGCGAAG[C/G]CTGCAGTGTCGGTGGGGCCAGTGGC
- Phenotype
-
MIM: 612209
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MSGN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34069439] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MSGN1
- Gene Name
- mesogenin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001105569.1 |
242 |
Missense Mutation |
GCC,GGC |
A81G |
NP_001099039.1 |
View Full Product Details