Product Details
- SNP ID
-
rs200799044
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:17817019 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAGCCTCTCACCAAGATCCAGAC[A/G]CTCAAGTACACCATCAAGTACATCG
- Phenotype
-
MIM: 612209
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MSGN1
PubMed Links
Gene Details
- Gene
- MSGN1
- Gene Name
- mesogenin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001105569.1 |
501 |
Silent Mutation |
ACA,ACG |
T167T |
NP_001099039.1 |
View Full Product Details