Product Details

SNP ID
rs199691868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27150264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTGGACGTGCTGGTGCTCGCC[A/G]CCAGCTACATAGCCCACCTCACCCG
Phenotype
MIM: 609635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCF23 PubMed Links

Gene Details

Gene
TCF23
Gene Name
transcription factor 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175769.2 421 Missense Mutation ACC,GCC T122A NP_786951.1
XM_005264159.4 421 Missense Mutation ACC,GCC T122A XP_005264216.1

View Full Product Details