Product Details
- SNP ID
-
rs200577403
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:224770237 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCATTCATGACTGTGGAGAGTT[C/T]GCTGAGCATGTCCTTGTAGTGGGAC
- Phenotype
-
MIM: 611518
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DOCK10
PubMed Links
Gene Details
- Gene
- DOCK10
- Gene Name
- dedicator of cytokinesis 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290263.1 |
6733 |
Missense Mutation |
AAA,GAA |
K2134E |
NP_001277192.1 |
NM_014689.2 |
6733 |
Missense Mutation |
AAA,GAA |
K2140E |
NP_055504.2 |
XM_005246673.4 |
6733 |
Missense Mutation |
AAA,GAA |
K2153E |
XP_005246730.1 |
XM_006712617.3 |
6733 |
Missense Mutation |
AAA,GAA |
K2153E |
XP_006712680.1 |
XM_017004441.1 |
6733 |
Missense Mutation |
AAA,GAA |
K2153E |
XP_016859930.1 |
XM_017004442.1 |
6733 |
Missense Mutation |
AAA,GAA |
K2144E |
XP_016859931.1 |
XM_017004443.1 |
6733 |
Missense Mutation |
AAA,GAA |
K2140E |
XP_016859932.1 |
XM_017004444.1 |
6733 |
Missense Mutation |
AAA,GAA |
K2138E |
XP_016859933.1 |
XM_017004445.1 |
6733 |
Missense Mutation |
AAA,GAA |
K2134E |
XP_016859934.1 |
XM_017004446.1 |
6733 |
Missense Mutation |
AAA,GAA |
K2104E |
XP_016859935.1 |
XM_017004447.1 |
6733 |
Missense Mutation |
AAA,GAA |
K2144E |
XP_016859936.1 |
View Full Product Details