Product Details

SNP ID
rs200561796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219210764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGATGGTGCGGGCAATGGCGA[C/T]GCGCTGCTTCTCCCCGCCGCTCAGC
Phenotype
MIM: 605452 MIM: 612204 MIM: 613474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCB6 PubMed Links

Gene Details

Gene
ABCB6
Gene Name
ATP binding cassette subfamily B member 6 (Langereis blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005689.2 2520 Missense Mutation ATC,GTC I735V NP_005680.1
Gene
ATG9A
Gene Name
autophagy related 9A
There are no transcripts associated with this gene.

Gene
ZFAND2B
Gene Name
zinc finger AN1-type containing 2B
There are no transcripts associated with this gene.

View Full Product Details