Product Details

SNP ID
rs200405363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231925452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATCCCAGGCCGCTCATGGAGCC[G/T]ATTCGGTCCAGCTTGAGGCCGAAGC
Phenotype
MIM: 600296
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NPPC PubMed Links

Gene Details

Gene
NPPC
Gene Name
natriuretic peptide C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024409.3 492 Silent Mutation ATA,ATC I118I NP_077720.1
XM_011511245.2 492 Silent Mutation ATA,ATC I118I XP_011509547.1

View Full Product Details