Product Details

SNP ID
rs200085375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86789426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCACAGGCGAAGTCCAGCCCCCT[C/G]GTGTGCACTGACGACACCAAAGACG
Phenotype
MIM: 186910
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD8A PubMed Links

Gene Details

Gene
CD8A
Gene Name
CD8a molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145873.1 1553 Silent Mutation ACC,ACG T174T NP_001139345.1
NM_001768.6 1553 Silent Mutation ACC,ACG T174T NP_001759.3
NM_171827.3 1553 Intron NP_741969.1

View Full Product Details