Product Details

SNP ID
rs200465701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101355269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACACACCTTGAAAACATGGCTT[G/T]CTTGGCAAATTCTACTTCTTCTGGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CREG2 PubMed Links

Gene Details

Gene
CREG2
Gene Name
cellular repressor of E1A stimulated genes 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153836.3 755 Missense Mutation NP_722578.1
XM_011510777.2 755 Intron XP_011509079.1
XM_017003565.1 755 Intron XP_016859054.1

View Full Product Details