Product Details

SNP ID
rs199921991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38931156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCACAGGCCGGGCCTGCCCAAG[C/G]ACAAGGAAGTCCTTTTCGTTCTATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF33 PubMed Links

Gene Details

Gene
ARHGEF33
Gene Name
Rho guanine nucleotide exchange factor 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145451.2 495 Missense Mutation GCA,GGA A137G NP_001138923.2

View Full Product Details