Product Details

SNP ID
rs201642274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169510231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCTTGACTGCCCGAGACTCGC[A/C]ATTTCTGCCCGTGAATTTGTGTCTG
Phenotype
MIM: 603650 MIM: 607701
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BBS5 PubMed Links

Gene Details

Gene
BBS5
Gene Name
Bardet-Biedl syndrome 5
There are no transcripts associated with this gene.

Gene
KLHL41
Gene Name
kelch like family member 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006063.2 530 Silent Mutation GCA,GCC A151A NP_006054.2

View Full Product Details