Product Details

SNP ID
rs201854875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96861320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGGAGCCGGGCATCGTAGAGG[A/T]AGGACCCGGGCTGTTCCGCAGGCAG
Phenotype
MIM: 604462
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD39 PubMed Links

Gene Details

Gene
ANKRD39
Gene Name
ankyrin repeat domain 39
There are no transcripts associated with this gene.

Gene
SEMA4C
Gene Name
semaphorin 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017789.4 1968 Missense Mutation TAC,TTC Y603F NP_060259.4
XM_006712606.3 1968 Intron XP_006712669.2
XM_011511378.2 1968 Missense Mutation TAC,TTC Y603F XP_011509680.1
XM_011511379.2 1968 Missense Mutation TAC,TTC Y603F XP_011509681.1
XM_011511380.1 1968 Missense Mutation TAC,TTC Y603F XP_011509682.1
XM_011511381.1 1968 Missense Mutation TAC,TTC Y603F XP_011509683.1
XM_011511382.2 1968 Missense Mutation TAC,TTC Y603F XP_011509684.1
XM_011511383.1 1968 Missense Mutation TAC,TTC Y603F XP_011509685.1
XM_017004393.1 1968 Missense Mutation TAC,TTC Y645F XP_016859882.1
XM_017004394.1 1968 Missense Mutation TAC,TTC Y462F XP_016859883.1

View Full Product Details