Product Details

SNP ID
rs202107782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206004649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGCTGCTGGGGGTCACGGTGG[C/T]GAAGGCAGGCTGTGTGGTCTCTGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INO80D PubMed Links

Gene Details

Gene
INO80D
Gene Name
INO80 complex subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017759.4 3040 Missense Mutation NP_060229.3
XM_011511369.2 3040 Missense Mutation XP_011509671.1
XM_011511370.2 3040 Missense Mutation XP_011509672.1
XM_011511371.2 3040 Missense Mutation XP_011509673.1
XM_011511373.2 3040 Missense Mutation XP_011509675.1
XM_011511375.2 3040 Missense Mutation XP_011509677.1
XM_011511376.2 3040 Missense Mutation XP_011509678.1

View Full Product Details