Product Details

SNP ID
rs201870543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:53853515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATTTGTTTCTGTTGACTTCGAG[A/G]TTCCCTTTTCCCATGGCTCGAAGTC
Phenotype
MIM: 606704
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR75 PubMed Links

Gene Details

Gene
GPR75
Gene Name
G protein-coupled receptor 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006794.3 1557 Silent Mutation NP_006785.1
Gene
GPR75-ASB3
Gene Name
GPR75-ASB3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164165.1 1557 Intron NP_001157637.1
Gene
MIR3682
Gene Name
microRNA 3682
There are no transcripts associated with this gene.

View Full Product Details