Product Details

SNP ID
rs201648655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74527043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGCTGTTGGCTGAGGGGTCACC[A/G]GGCCAGAGCTGGGAAACTGAGGTGA
Phenotype
MIM: 602434 MIM: 606441 MIM: 607163 MIM: 604240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AUP1 PubMed Links

Gene Details

Gene
AUP1
Gene Name
ancient ubiquitous protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181575.4 1307 Missense Mutation CCG,CTG P365L NP_853553.1
XM_005264392.3 1307 Missense Mutation CCG,CTG P422L XP_005264449.1
Gene
DQX1
Gene Name
DEAQ-box RNA dependent ATPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133637.2 1307 Intron NP_598376.2
XM_011532644.1 1307 Intron XP_011530946.1
XM_011532645.1 1307 Intron XP_011530947.1
XM_011532646.2 1307 Intron XP_011530948.1
XM_017003520.1 1307 Intron XP_016859009.1
Gene
HTRA2
Gene Name
HtrA serine peptidase 2
There are no transcripts associated with this gene.

Gene
LOXL3
Gene Name
lysyl oxidase like 3
There are no transcripts associated with this gene.

Gene
TLX2
Gene Name
T-cell leukemia homeobox 2
There are no transcripts associated with this gene.

View Full Product Details