Product Details

SNP ID
rs201636455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74834632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCACGGAGCTCAACCATGAC[C/G]AAGTGCAGAAGGTAAGTCAGCGCGG
Phenotype
MIM: 601125
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HK2 PubMed Links

Gene Details

Gene
HK2
Gene Name
hexokinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000189.4 1978 Missense Mutation CAA,GAA Q18E NP_000180.2
XM_005264280.1 1978 Intron XP_005264337.1
XM_011532807.1 1978 Intron XP_011531109.1
XM_017003945.1 1978 Intron XP_016859434.1

View Full Product Details