Product Details

SNP ID
rs201663885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64636040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGAGCCAGAGTTTTGAGGAGGT[C/T]GTCGGCAGACACAGGGGCCATTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERTAD2 PubMed Links

Gene Details

Gene
SERTAD2
Gene Name
SERTA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014755.2 1183 Missense Mutation AAC,GAC N278D NP_055570.1
XM_005264669.2 1183 Missense Mutation AAC,GAC N278D XP_005264726.1
XM_011533204.1 1183 Missense Mutation AAC,GAC N285D XP_011531506.1
XM_011533205.2 1183 Missense Mutation AAC,GAC N278D XP_011531507.1

View Full Product Details