Product Details
- SNP ID
-
rs201858817
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:158172199 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAATATCTCTTTAGCATAAAGTG[G/T]TCTATGAAGTCCAGCTTCTCGAAGT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC148
PubMed Links
Gene Details
- Gene
- CCDC148
- Gene Name
- coiled-coil domain containing 148
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301684.1 |
1939 |
Missense Mutation |
ACA,CCA |
T418P |
NP_001288613.1 |
NM_001301685.1 |
1939 |
Intron |
|
|
NP_001288614.1 |
NM_138803.3 |
1939 |
Missense Mutation |
ACA,CCA |
T564P |
NP_620158.3 |
XM_005246319.3 |
1939 |
Missense Mutation |
ACA,CCA |
T585P |
XP_005246376.1 |
XM_011510638.2 |
1939 |
Missense Mutation |
ACA,CCA |
T598P |
XP_011508940.1 |
XM_017003392.1 |
1939 |
Missense Mutation |
ACA,CCA |
T577P |
XP_016858881.1 |
XM_017003393.1 |
1939 |
Missense Mutation |
ACA,CCA |
T563P |
XP_016858882.1 |
XM_017003394.1 |
1939 |
Missense Mutation |
ACA,CCA |
T524P |
XP_016858883.1 |
XM_017003395.1 |
1939 |
Missense Mutation |
ACA,CCA |
T491P |
XP_016858884.1 |
XM_017003396.1 |
1939 |
Missense Mutation |
ACA,CCA |
T478P |
XP_016858885.1 |
XM_017003397.1 |
1939 |
Missense Mutation |
ACA,CCA |
T418P |
XP_016858886.1 |
XM_017003398.1 |
1939 |
Missense Mutation |
ACA,CCA |
T351P |
XP_016858887.1 |
- Gene
- CCDC148-AS1
- Gene Name
- CCDC148 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details