Product Details

SNP ID
rs199536811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:197974401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCATAAATAAAGTTCATCTTAAG[A/G]CAAATCATGTGGTCAAGAGAGATGT
Phenotype
MIM: 609380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMLN PubMed Links

Gene Details

Gene
LMLN
Gene Name
leishmanolysin like peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136049.2 249 Missense Mutation ACA,GCA T82A NP_001129521.2
NM_033029.3 249 Missense Mutation ACA,GCA T82A NP_149018.2
XM_011513270.1 249 Missense Mutation ACA,GCA T82A XP_011511572.1
XM_011513271.1 249 Missense Mutation ACA,GCA T82A XP_011511573.1
XM_017007428.1 249 Missense Mutation ACA,GCA T82A XP_016862917.1

View Full Product Details