Product Details

SNP ID
rs202010878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52054237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCAGCTCTCGGTCCGACTCGC[C/T]GTCGGAGCAGTCAGAGCTGATGCGC
Phenotype
MIM: 602749
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP7 PubMed Links

Gene Details

Gene
DUSP7
Gene Name
dual specificity phosphatase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001947.3 734 Missense Mutation AGC,GGC S219G NP_001938.2
Gene
LINC00696
Gene Name
long intergenic non-protein coding RNA 696
There are no transcripts associated with this gene.

View Full Product Details