Product Details

SNP ID
rs201543433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49909016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGAGCCCGCACACATTCCCATC[A/G]ATAGGTGAGGGGCGTGAGAATGAAG
Phenotype
MIM: 611464 MIM: 600168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MON1A PubMed Links

Gene Details

Gene
MON1A
Gene Name
MON1 homolog A, secretory trafficking associated
There are no transcripts associated with this gene.

Gene
MST1R
Gene Name
macrophage stimulating 1 receptor
There are no transcripts associated with this gene.

View Full Product Details