Product Details

SNP ID
rs201544209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98087298 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGTCTATCTCATCACCATGGT[G/T]GGCAACCTTGGACTGATAGTTCTAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR5AC2 PubMed Links

Gene Details

Gene
OR5AC2
Gene Name
olfactory receptor family 5 subfamily AC member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054106.1 126 Silent Mutation GTG,GTT V42V NP_473447.1

View Full Product Details