Product Details

SNP ID
rs200939350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51712790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCTTTGAGGGTGTGGTGCGAGC[C/T]CTGCTGCAGAAGCCCAGTGCCCGCG
Phenotype
MIM: 604099
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRM2 PubMed Links

Gene Details

Gene
GRM2
Gene Name
glutamate metabotropic receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000839.3 1485 Silent Mutation GCC,GCT A256A NP_000830.2
NM_001130063.1 1485 Intron NP_001123535.1
XM_011533635.1 1485 Silent Mutation GCC,GCT A256A XP_011531937.1
XM_011533636.1 1485 Intron XP_011531938.1
XM_011533637.1 1485 Silent Mutation GCC,GCT A62A XP_011531939.1
XM_011533638.2 1485 Silent Mutation GCC,GCT A62A XP_011531940.1
XM_011533639.2 1485 Silent Mutation GCC,GCT A62A XP_011531941.1
XM_011533640.2 1485 Silent Mutation GCC,GCT A62A XP_011531942.1
XM_011533641.2 1485 Silent Mutation GCC,GCT A62A XP_011531943.1
XM_011533642.1 1485 Silent Mutation GCC,GCT A256A XP_011531944.1
XM_011533643.1 1485 Intron XP_011531945.1
XM_011533644.1 1485 Intron XP_011531946.1
XM_011533645.1 1485 Intron XP_011531947.1
XM_011533646.2 1485 Intron XP_011531948.1
XM_011533647.1 1485 Intron XP_011531949.1
XM_017006271.1 1485 Silent Mutation GCC,GCT A256A XP_016861760.1
Gene
TEX264
Gene Name
testis expressed 264
There are no transcripts associated with this gene.

View Full Product Details