Product Details

SNP ID
rs200924801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196198827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCACAAATTGTTAAATACCAGTT[G/T]CTGGCACAGCCCTGGTCGAAGGGGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZDHHC19 PubMed Links

Gene Details

Gene
ZDHHC19
Gene Name
zinc finger DHHC-type containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039617.1 827 Missense Mutation AGA,AGC R245S NP_001034706.1
XM_005269279.2 827 Intron XP_005269336.1
XM_006713493.2 827 Missense Mutation AGA,AGC R243S XP_006713556.1
XM_006713494.2 827 Missense Mutation AGA,AGC R219S XP_006713557.1
XM_011512417.1 827 Missense Mutation AGA,AGC R171S XP_011510719.1

View Full Product Details