Product Details
- SNP ID
-
rs201236076
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:65356600 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTGGCCCTGCGCTCGGGCGAGCC[C/T]CCTCTCCTGCGCTCGGGGGACCTCC
- Phenotype
-
MIM: 602625
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAGI1
PubMed Links
Gene Details
- Gene
- MAGI1
- Gene Name
- membrane associated guanylate kinase, WW and PDZ domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033057.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1389G |
NP_001028229.1 |
NM_004742.2 |
4874 |
Intron |
|
|
NP_004733.2 |
NM_015520.1 |
4874 |
UTR 3 |
|
|
NP_056335.1 |
XM_005265563.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1419G |
XP_005265620.1 |
XM_005265564.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1418G |
XP_005265621.1 |
XM_005265565.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1408G |
XP_005265622.1 |
XM_005265566.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1406G |
XP_005265623.1 |
XM_005265568.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1405G |
XP_005265625.1 |
XM_005265570.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1352G |
XP_005265627.1 |
XM_005265571.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1322G |
XP_005265628.1 |
XM_005265574.3 |
4874 |
UTR 3 |
|
|
XP_005265631.1 |
XM_006713407.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1420G |
XP_006713470.1 |
XM_006713408.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1419G |
XP_006713471.1 |
XM_006713409.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1419G |
XP_006713472.1 |
XM_006713410.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1406G |
XP_006713473.1 |
XM_006713411.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1392G |
XP_006713474.1 |
XM_006713412.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1390G |
XP_006713475.1 |
XM_006713414.3 |
4874 |
UTR 3 |
|
|
XP_006713477.1 |
XM_011534236.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1407G |
XP_011532538.1 |
XM_011534240.2 |
4874 |
Silent Mutation |
GGA,GGG |
G701G |
XP_011532542.1 |
XM_011534241.2 |
4874 |
Silent Mutation |
GGA,GGG |
G673G |
XP_011532543.1 |
XM_017007497.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1418G |
XP_016862986.1 |
XM_017007498.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1417G |
XP_016862987.1 |
XM_017007499.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1385G |
XP_016862988.1 |
XM_017007500.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1382G |
XP_016862989.1 |
XM_017007501.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1379G |
XP_016862990.1 |
XM_017007502.1 |
4874 |
UTR 3 |
|
|
XP_016862991.1 |
XM_017007503.1 |
4874 |
UTR 3 |
|
|
XP_016862992.1 |
XM_017007504.1 |
4874 |
UTR 3 |
|
|
XP_016862993.1 |
XM_017007505.1 |
4874 |
UTR 3 |
|
|
XP_016862994.1 |
XM_017007506.1 |
4874 |
UTR 3 |
|
|
XP_016862995.1 |
XM_017007507.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1151G |
XP_016862996.1 |
XM_017007508.1 |
4874 |
Silent Mutation |
GGA,GGG |
G1095G |
XP_016862997.1 |
View Full Product Details