Product Details
- SNP ID
-
rs201289437
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:52435623 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTTCTACGTGGCCTCCACCTCCC[A/G]GGGCGTCCGATTGTGCTCGGCATGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEMA3G
PubMed Links
Gene Details
- Gene
- SEMA3G
- Gene Name
- semaphorin 3G
There are no transcripts associated with this gene.
View Full Product Details