Product Details

SNP ID
rs200913034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53495229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGGCAGCAGCAAGCGGACCACGC[G/T]AACGGTGAGCAGCCAGAGCCCGGGC
Phenotype
MIM: 114206
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CACNA1D PubMed Links

Gene Details

Gene
CACNA1D
Gene Name
calcium voltage-gated channel subunit alpha1 D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000720.3 63 Silent Mutation GCG,GCT A21A NP_000711.1
NM_001128839.2 63 Silent Mutation GCG,GCT A21A NP_001122311.1
NM_001128840.2 63 Silent Mutation GCG,GCT A21A NP_001122312.1
XM_005265448.3 63 Silent Mutation GCG,GCT A21A XP_005265505.1
XM_011534094.2 63 Intron XP_011532396.1
XM_011534096.2 63 Intron XP_011532398.1
XM_011534097.2 63 Intron XP_011532399.1
XM_011534099.2 63 Intron XP_011532401.1
XM_011534100.2 63 Intron XP_011532402.1
XM_017007137.1 63 Intron XP_016862626.1
XM_017007138.1 63 Intron XP_016862627.1
XM_017007139.1 63 Intron XP_016862628.1
XM_017007140.1 63 Intron XP_016862629.1
XM_017007141.1 63 Intron XP_016862630.1
XM_017007142.1 63 Intron XP_016862631.1
XM_017007143.1 63 Intron XP_016862632.1
XM_017007144.1 63 Intron XP_016862633.1
XM_017007145.1 63 Intron XP_016862634.1

View Full Product Details