Product Details

SNP ID
rs200997728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49276781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAATAACTAACTCCTTCAAAG[G/T]CCCGGTCAATGGCTGCAGTCAGTTC
Phenotype
MIM: 603486
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C3orf62 PubMed Links

Gene Details

Gene
C3orf62
Gene Name
chromosome 3 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198562.2 295 Missense Mutation NP_940964.1
Gene
MIR4271
Gene Name
microRNA 4271
There are no transcripts associated with this gene.

Gene
USP4
Gene Name
ubiquitin specific peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251877.1 295 Intron NP_001238806.1
NM_003363.3 295 Intron NP_003354.2
NM_199443.2 295 Intron NP_955475.1

View Full Product Details