Product Details
- SNP ID
-
rs201705102
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:47501672 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTTTAGTTCCCAGCACACGGTG[A/G]CTCTGCAGTAGTGAATGAAGTCTAG
- Phenotype
-
MIM: 615020
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ELP6
PubMed Links
Gene Details
- Gene
- ELP6
- Gene Name
- elongator acetyltransferase complex subunit 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001031703.2 |
601 |
Missense Mutation |
GCC,GTC |
A168V |
NP_001026873.2 |
XM_005265241.3 |
601 |
Missense Mutation |
GCC,GTC |
A119V |
XP_005265298.1 |
XM_011533851.2 |
601 |
Missense Mutation |
GCC,GTC |
A168V |
XP_011532153.1 |
XM_011533852.2 |
601 |
Missense Mutation |
GCC,GTC |
A165V |
XP_011532154.1 |
XM_011533853.2 |
601 |
Missense Mutation |
GCC,GTC |
A151V |
XP_011532155.1 |
XM_011533854.2 |
601 |
Missense Mutation |
GCC,GTC |
A119V |
XP_011532156.1 |
XM_011533856.2 |
601 |
Missense Mutation |
GCC,GTC |
A119V |
XP_011532158.1 |
XM_011533857.2 |
601 |
Missense Mutation |
GCC,GTC |
A119V |
XP_011532159.1 |
XM_017006669.1 |
601 |
Missense Mutation |
GCC,GTC |
A168V |
XP_016862158.1 |
XM_017006670.1 |
601 |
Missense Mutation |
GCC,GTC |
A95V |
XP_016862159.1 |
View Full Product Details