Product Details

SNP ID
rs200709234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33152770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTTCTCCATCTCGATCTGCTGG[C/T]GGTAGTGCCGAGCCTGCCGCTGGCC
Phenotype
MIM: 605497
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRTAP PubMed Links

Gene Details

Gene
CRTAP
Gene Name
cartilage associated protein
There are no transcripts associated with this gene.

Gene
SUSD5
Gene Name
sushi domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015551.1 2280 Missense Mutation CAC,CGC H621R NP_056366.1
XM_005265034.3 2280 Missense Mutation CAC,CGC H558R XP_005265091.1
XM_017006137.1 2280 Missense Mutation CAC,CGC H511R XP_016861626.1

View Full Product Details