Product Details

SNP ID
rs201362782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49909271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTACCCAGGCCAGGAGGTTCTC[A/G]TTGGGGCCCGTGTAGTAAATGGTCT
Phenotype
MIM: 611464 MIM: 600168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MON1A PubMed Links

Gene Details

Gene
MON1A
Gene Name
MON1 homolog A, secretory trafficking associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142501.1 1926 Silent Mutation NP_001135973.1
NM_032355.3 1926 Silent Mutation NP_115731.2
XM_006713345.3 1926 Silent Mutation XP_006713408.1
XM_011534160.1 1926 Silent Mutation XP_011532462.1
Gene
MST1R
Gene Name
macrophage stimulating 1 receptor
There are no transcripts associated with this gene.

View Full Product Details