Product Details

SNP ID
rs200367729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121772619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCTTCTCTGTGCTGCTGGGCT[C/T]GCTCTTCTAGGGCCCTGCCCATAAA
Phenotype
MIM: 609237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQCB1 PubMed Links

Gene Details

Gene
IQCB1
Gene Name
IQ motif containing B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023570.3 1404 Intron NP_001018864.2
NM_001023571.3 1404 Intron NP_001018865.2
NM_001319107.1 1404 Intron NP_001306036.1
XM_005247911.3 1404 Intron XP_005247968.1
XM_005247912.2 1404 Intron XP_005247969.1
XM_011513335.2 1404 Intron XP_011511637.1
XM_017007537.1 1404 Intron XP_016863026.1
XM_017007538.1 1404 Missense Mutation CAA,CGA Q318R XP_016863027.1
XM_017007539.1 1404 Intron XP_016863028.1

View Full Product Details