Product Details

SNP ID
rs200188637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53865028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTGTGCGATGGTACCTGTGGCA[A/C]GAGCGAGGGCAGTCCCAGTGAGAAC
Phenotype
MIM: 605458
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACTR8 PubMed Links

Gene Details

Gene
ACTR8
Gene Name
ARP8 actin-related protein 8 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022899.4 1137 Intron NP_075050.3
XM_005265587.4 1137 Intron XP_005265644.1
XM_011534249.2 1137 Intron XP_011532551.1
Gene
IL17RB
Gene Name
interleukin 17 receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018725.3 1137 Missense Mutation AAG,ACG K410T NP_061195.2
XM_005265310.4 1137 Missense Mutation AAG,ACG K439T XP_005265367.1
XM_005265311.4 1137 Missense Mutation AAG,ACG K423T XP_005265368.1
XM_005265312.4 1137 Missense Mutation AAG,ACG K394T XP_005265369.1
XM_011533941.2 1137 UTR 3 XP_011532243.1
XM_017006804.1 1137 Missense Mutation AAG,ACG K322T XP_016862293.1
XM_017006805.1 1137 Missense Mutation AAG,ACG K306T XP_016862294.1
XM_017006806.1 1137 Missense Mutation AAG,ACG K293T XP_016862295.1
XM_017006807.1 1137 UTR 3 XP_016862296.1

View Full Product Details