Product Details
- SNP ID
-
rs200547630
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:152835974 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTCTACATCTTGGTATTCATCAT[C/T]GGCTTCCTGGGCAACAGCGTGGCCA
- Phenotype
-
MIM: 601167
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
P2RY1
PubMed Links
Gene Details
- Gene
- P2RY1
- Gene Name
- purinergic receptor P2Y1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002563.4 |
1282 |
Silent Mutation |
ATC,ATT |
I64I |
NP_002554.1 |
View Full Product Details