Product Details

SNP ID
rs200463539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:25719630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAATCAGCATAGGAGTGAAGAC[C/T]GTTATCTGTTAGAGGGAAAAAAAAA
Phenotype
MIM: 610661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NGLY1 PubMed Links

Gene Details

Gene
NGLY1
Gene Name
N-glycanase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145293.1 1725 Missense Mutation AGT,GGT S581G NP_001138765.1
NM_001145294.1 1725 Missense Mutation AGT,GGT S557G NP_001138766.1
NM_001145295.1 1725 Silent Mutation ACA,ACG T539T NP_001138767.1
NM_018297.3 1725 Missense Mutation AGT,GGT S599G NP_060767.2
XM_005265316.1 1725 Missense Mutation AGT,GGT S544G XP_005265373.1
XM_005265317.1 1725 Silent Mutation ACA,ACG T484T XP_005265374.1
XM_011533944.1 1725 Missense Mutation AGT,GGT S522G XP_011532246.1
XM_011533945.2 1725 Intron XP_011532247.1
XM_017006839.1 1725 Intron XP_016862328.1

View Full Product Details