Product Details

SNP ID
rs199733019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52707696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTTCTGGCCCAGCTGACACTT[C/G]CTCCATGGCCCATCTATCGCCGGCA
Phenotype
MIM: 601959 MIM: 610358
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLT8D1 PubMed Links

Gene Details

Gene
GLT8D1
Gene Name
glycosyltransferase 8 domain containing 1
There are no transcripts associated with this gene.

Gene
NEK4
Gene Name
NIMA related kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193533.1 599 Intron NP_001180462.1
NM_003157.4 599 Intron NP_003148.2
XM_006713310.2 599 Intron XP_006713373.1
XM_011534039.2 599 Intron XP_011532341.1
XM_011534040.2 599 Intron XP_011532342.1
XM_017007085.1 599 Intron XP_016862574.1
XM_017007086.1 599 Intron XP_016862575.1
Gene
SPCS1
Gene Name
signal peptidase complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014041.3 599 Missense Mutation CCT,GCT P132A NP_054760.3

View Full Product Details