Product Details

SNP ID
rs200314495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51986499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGAGGGCAGGCTTTGCCCTGG[A/G]TGAGGGTGAGCAGGTTGGCAAGCCA
Phenotype
MIM: 104620 MIM: 601832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD14A PubMed Links

Gene Details

Gene
ABHD14A
Gene Name
abhydrolase domain containing 14A
There are no transcripts associated with this gene.

Gene
ABHD14A-ACY1
Gene Name
ABHD14A-ACY1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316331.1 1046 Missense Mutation GAT,GGT D264G NP_001303260.1
Gene
ACY1
Gene Name
aminoacylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000666.2 1046 Missense Mutation GAT,GGT D174G NP_000657.1
NM_001198895.1 1046 Missense Mutation GAT,GGT D174G NP_001185824.1
NM_001198896.1 1046 Intron NP_001185825.1
NM_001198897.1 1046 Missense Mutation GAT,GGT D174G NP_001185826.1
NM_001198898.1 1046 Missense Mutation GAT,GGT D139G NP_001185827.1
Gene
RPL29
Gene Name
ribosomal protein L29
There are no transcripts associated with this gene.

View Full Product Details