Product Details

SNP ID
rs200689182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183320081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGGTTTGCCAGAGTCCCATGGT[C/G]CAGTTTTCCATTTCATTTAAGTCCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLDN22 PubMed Links

Gene Details

Gene
CLDN22
Gene Name
claudin 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111319.1 694 Missense Mutation TGC,TGG C46W NP_001104789.1
Gene
CLDN24
Gene Name
claudin 24
There are no transcripts associated with this gene.

Gene
WWC2
Gene Name
WW and C2 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024949.5 694 UTR 3 NP_079225.5
XM_011532269.2 694 Intron XP_011530571.1

View Full Product Details