Product Details
- SNP ID
-
rs200775951
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:169401696 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGCACTTTCACTGCTGGGATTG[G/T]CATCTTCATCTGCCACATCTGTCCC
- Phenotype
-
MIM: 604588
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NEK1
PubMed Links
Gene Details
- Gene
- NEK1
- Gene Name
- NIMA related kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199397.1 |
4035 |
Missense Mutation |
GAC,GCC |
D1180A |
NP_001186326.1 |
NM_001199398.1 |
4035 |
Missense Mutation |
GAC,GCC |
D1136A |
NP_001186327.1 |
NM_001199399.1 |
4035 |
Missense Mutation |
GAC,GCC |
D1083A |
NP_001186328.1 |
NM_001199400.1 |
4035 |
Missense Mutation |
GAC,GCC |
D1108A |
NP_001186329.1 |
NM_012224.2 |
4035 |
Missense Mutation |
GAC,GCC |
D1152A |
NP_036356.1 |
XM_006714228.1 |
4035 |
Missense Mutation |
GAC,GCC |
D1121A |
XP_006714291.1 |
XM_011532003.1 |
4035 |
Missense Mutation |
GAC,GCC |
D1152A |
XP_011530305.1 |
XM_011532004.1 |
4035 |
Missense Mutation |
GAC,GCC |
D1108A |
XP_011530306.1 |
XM_011532005.2 |
4035 |
Intron |
|
|
XP_011530307.1 |
XM_017008249.1 |
4035 |
Missense Mutation |
GAC,GCC |
D973A |
XP_016863738.1 |
XM_017008250.1 |
4035 |
Missense Mutation |
GAC,GCC |
D973A |
XP_016863739.1 |
XM_017008251.1 |
4035 |
Missense Mutation |
GAC,GCC |
D945A |
XP_016863740.1 |
XM_017008252.1 |
4035 |
Missense Mutation |
GAC,GCC |
D945A |
XP_016863741.1 |
XM_017008253.1 |
4035 |
Missense Mutation |
GAC,GCC |
D796A |
XP_016863742.1 |
XM_017008254.1 |
4035 |
Missense Mutation |
GAC,GCC |
D728A |
XP_016863743.1 |
View Full Product Details