Product Details
- SNP ID
-
rs200527704
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:16508607 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGCCAGTGCTGTTTGCATTGTT[C/T]CCAGCGCTGCTGTTGGAAGTGCTGC
- Phenotype
-
MIM: 603450
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LDB2
PubMed Links
Gene Details
- Gene
- LDB2
- Gene Name
- LIM domain binding 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130834.2 |
1173 |
Silent Mutation |
GGA,GGG |
G273G |
NP_001124306.1 |
NM_001290.4 |
1173 |
Silent Mutation |
GGA,GGG |
G273G |
NP_001281.1 |
NM_001304434.1 |
1173 |
Silent Mutation |
GGA,GGG |
G273G |
NP_001291363.1 |
NM_001304435.1 |
1173 |
Silent Mutation |
GGA,GGG |
G273G |
NP_001291364.1 |
XM_005248197.3 |
1173 |
Silent Mutation |
GGA,GGG |
G272G |
XP_005248254.1 |
XM_005248201.2 |
1173 |
Silent Mutation |
GGA,GGG |
G272G |
XP_005248258.1 |
XM_005248202.2 |
1173 |
Silent Mutation |
GGA,GGG |
G273G |
XP_005248259.1 |
XM_006713975.3 |
1173 |
Silent Mutation |
GGA,GGG |
G311G |
XP_006714038.1 |
XM_006713976.3 |
1173 |
Silent Mutation |
GGA,GGG |
G310G |
XP_006714039.1 |
XM_006713977.3 |
1173 |
Silent Mutation |
GGA,GGG |
G311G |
XP_006714040.1 |
XM_006713978.3 |
1173 |
Silent Mutation |
GGA,GGG |
G311G |
XP_006714041.1 |
XM_006713979.3 |
1173 |
Silent Mutation |
GGA,GGG |
G311G |
XP_006714042.1 |
XM_006713980.3 |
1173 |
Silent Mutation |
GGA,GGG |
G310G |
XP_006714043.1 |
XM_006713981.3 |
1173 |
Silent Mutation |
GGA,GGG |
G311G |
XP_006714044.1 |
XM_006713982.3 |
1173 |
Silent Mutation |
GGA,GGG |
G311G |
XP_006714045.1 |
XM_006713983.3 |
1173 |
Silent Mutation |
GGA,GGG |
G311G |
XP_006714046.1 |
XM_011513905.2 |
1173 |
Silent Mutation |
GGA,GGG |
G198G |
XP_011512207.1 |
XM_017008812.1 |
1173 |
Silent Mutation |
GGA,GGG |
G310G |
XP_016864301.1 |
XM_017008813.1 |
1173 |
Silent Mutation |
GGA,GGG |
G272G |
XP_016864302.1 |
XM_017008814.1 |
1173 |
Silent Mutation |
GGA,GGG |
G310G |
XP_016864303.1 |
XM_017008815.1 |
1173 |
Silent Mutation |
GGA,GGG |
G310G |
XP_016864304.1 |
XM_017008816.1 |
1173 |
Silent Mutation |
GGA,GGG |
G310G |
XP_016864305.1 |
XM_017008817.1 |
1173 |
Silent Mutation |
GGA,GGG |
G272G |
XP_016864306.1 |
XM_017008818.1 |
1173 |
Silent Mutation |
GGA,GGG |
G230G |
XP_016864307.1 |
XM_017008819.1 |
1173 |
Silent Mutation |
GGA,GGG |
G272G |
XP_016864308.1 |
XM_017008820.1 |
1173 |
Silent Mutation |
GGA,GGG |
G273G |
XP_016864309.1 |
XM_017008821.1 |
1173 |
Silent Mutation |
GGA,GGG |
G272G |
XP_016864310.1 |
XM_017008822.1 |
1173 |
Silent Mutation |
GGA,GGG |
G149G |
XP_016864311.1 |
View Full Product Details