Product Details

SNP ID
rs200710438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:169401668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATGCACCTGAGTGCCATTCTTC[A/G]TTCAGGGCACTTTCACTGCTGGGAT
Phenotype
MIM: 604588
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEK1 PubMed Links

Gene Details

Gene
NEK1
Gene Name
NIMA related kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199397.1 4063 Silent Mutation NP_001186326.1
NM_001199398.1 4063 Silent Mutation NP_001186327.1
NM_001199399.1 4063 Silent Mutation NP_001186328.1
NM_001199400.1 4063 Silent Mutation NP_001186329.1
NM_012224.2 4063 Silent Mutation NP_036356.1
XM_006714228.1 4063 Silent Mutation XP_006714291.1
XM_011532003.1 4063 Silent Mutation XP_011530305.1
XM_011532004.1 4063 Silent Mutation XP_011530306.1
XM_011532005.2 4063 Intron XP_011530307.1
XM_017008249.1 4063 Silent Mutation XP_016863738.1
XM_017008250.1 4063 Silent Mutation XP_016863739.1
XM_017008251.1 4063 Silent Mutation XP_016863740.1
XM_017008252.1 4063 Silent Mutation XP_016863741.1
XM_017008253.1 4063 Silent Mutation XP_016863742.1
XM_017008254.1 4063 Silent Mutation XP_016863743.1

View Full Product Details