Product Details

SNP ID
rs200984113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:9783088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTTCGGCTGGGCTAACTCCTC[A/G]CTCAACCCCGTCATCTATGCCTTCA
Phenotype
MIM: 126453 MIM: 606142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRD5 PubMed Links

Gene Details

Gene
DRD5
Gene Name
dopamine receptor D5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000798.4 1455 Silent Mutation TCA,TCG S353S NP_000789.1
Gene
SLC2A9
Gene Name
solute carrier family 2 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001290.1 1455 Intron NP_001001290.1
NM_020041.2 1455 Intron NP_064425.2
XM_006713968.3 1455 Intron XP_006714031.1
XM_006713969.2 1455 Intron XP_006714032.1
XM_011513856.2 1455 Intron XP_011512158.1
XM_011513857.1 1455 Intron XP_011512159.1
XM_011513858.1 1455 Intron XP_011512160.1
XM_011513859.2 1455 Intron XP_011512161.1
XM_011513860.2 1455 Intron XP_011512162.1
XM_011513861.2 1455 Intron XP_011512163.1
XM_011513862.2 1455 Intron XP_011512164.1
XM_011513863.2 1455 Intron XP_011512165.1
XM_011513864.2 1455 Intron XP_011512166.1
XM_011513865.2 1455 Intron XP_011512167.1
XM_011513866.2 1455 Intron XP_011512168.1
XM_011513867.2 1455 Intron XP_011512169.1
XM_011513868.2 1455 Intron XP_011512170.1
XM_017008457.1 1455 Intron XP_016863946.1
XM_017008458.1 1455 Intron XP_016863947.1
XM_017008459.1 1455 Intron XP_016863948.1
XM_017008460.1 1455 Intron XP_016863949.1

View Full Product Details