Product Details
- SNP ID
-
rs200984268
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:153712556 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTTCCCAACGATACACCAACCT[C/T]GGATGCAGAATTCATGAAAGCTGAA
- Phenotype
-
MIM: 603028
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RNF175
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2289318] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RNF175
- Gene Name
- ring finger protein 175
- Gene
- TLR2
- Gene Name
- toll like receptor 2
There are no transcripts associated with this gene.
View Full Product Details