Product Details

SNP ID
rs200093346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:98476252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTAAATTGTCCTGCAACCACTT[C/T]TCAAACTGGGGCACACAGCCTTTCG
Phenotype
MIM: 613136
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSPAN5 PubMed Links

Gene Details

Gene
TSPAN5
Gene Name
tetraspanin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005723.3 1113 Silent Mutation GAA,GAG E226E NP_005714.2
XM_005262680.1 1113 Silent Mutation GAA,GAG E177E XP_005262737.1

View Full Product Details