Product Details

SNP ID
rs200752163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:44175208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTGACTTTGTCATGTTTCCTA[A/T]CTTCATGTTCTTGTTTAGGTGATAC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCTD8 PubMed Links

Gene Details

Gene
KCTD8
Gene Name
potassium channel tetramerization domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198353.2 1288 Missense Mutation GAT,GTT D335V NP_938167.1
XM_011513690.2 1288 Missense Mutation GAT,GTT D363V XP_011511992.1

View Full Product Details