Product Details

SNP ID
rs199759240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140389266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTGCGTACTGACTTTATCGG[C/T]CCATCTCCAGATCCTGTGCTCTCAT
Phenotype
MIM: 601858
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLGN PubMed Links

Gene Details

Gene
CLGN
Gene Name
calmegin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130675.1 1925 Silent Mutation GGA,GGG G597G NP_001124147.1
NM_004362.2 1925 Silent Mutation GGA,GGG G597G NP_004353.1
Gene
SCOC
Gene Name
short coiled-coil protein
There are no transcripts associated with this gene.

View Full Product Details