Product Details

SNP ID
rs199605036
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:125317232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCCAGCGTCCTCCAGGTGGCGG[C/T]GGCGGACGCGGACGAGGGCACCAAC
Phenotype
MIM: 612411
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAT4 PubMed Links

Gene Details

Gene
FAT4
Gene Name
FAT atypical cadherin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291285.1 821 Missense Mutation GCG,GTG A274V NP_001278214.1
NM_001291303.1 821 Missense Mutation GCG,GTG A274V NP_001278232.1
NM_024582.4 821 Missense Mutation GCG,GTG A274V NP_078858.4
XM_011532236.2 821 Missense Mutation GCG,GTG A274V XP_011530538.1
XM_011532237.2 821 Intron XP_011530539.1

View Full Product Details