Product Details

SNP ID
rs199642592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6709362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCACAACTCCGACACTCTCTCGT[A/C]GGCTTCGCCACTCGGGTGCTGCACG
Phenotype
MIM: 605695
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BLOC1S4 PubMed Links

Gene Details

Gene
BLOC1S4
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 4
There are no transcripts associated with this gene.

Gene
MRFAP1L1
Gene Name
Morf4 family associated protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203462.2 518 Missense Mutation GAC,TAC D90Y NP_982287.1
XM_005247932.1 518 Missense Mutation GAC,TAC D90Y XP_005247989.1

View Full Product Details