Product Details

SNP ID
rs201890938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:123401723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTATTTTGTCCTTAGACCAGAT[C/T]AAGGCCATAAGAGGCAGCAATGAAT
Phenotype
MIM: 602465
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPRY1 PubMed Links

Gene Details

Gene
SPRY1
Gene Name
sprouty RTK signaling antagonist 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258038.1 325 Silent Mutation ATC,ATT I44I NP_001244967.1
NM_001258039.1 325 Silent Mutation ATC,ATT I44I NP_001244968.1
NM_005841.2 325 Silent Mutation ATC,ATT I44I NP_005832.1
NM_199327.2 325 Silent Mutation ATC,ATT I44I NP_955359.1
XM_005262686.1 325 Silent Mutation ATC,ATT I44I XP_005262743.1
XM_017007635.1 325 Silent Mutation ATC,ATT I44I XP_016863124.1

View Full Product Details