Product Details

SNP ID
rs201445547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6576636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCAGTGGTGGAAGAGCTGGCCC[A/G]CGGCCAGCAGCGCCGGTTCATCGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAN2B2 PubMed Links

Gene Details

Gene
MAN2B2
Gene Name
mannosidase alpha class 2B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292038.1 234 Missense Mutation CAC,CGC H66R NP_001278967.1
NM_015274.2 234 Missense Mutation CAC,CGC H66R NP_056089.1

View Full Product Details