Product Details

SNP ID
rs201120262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88463539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTTTCCTTACATAGGTAAAGA[A/T]GATCCATCCCTTATTGAAGGACTAG
Phenotype
MIM: 608242
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HERC5 PubMed Links

Gene Details

Gene
HERC5
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016323.3 1362 Missense Mutation GAA,GAT E232D NP_057407.2
XM_011532022.2 1362 Missense Mutation GAA,GAT E383D XP_011530324.2

View Full Product Details